Anti-ANXA1 Mouse Monoclonal Antibody [clone: 686122]

Supplier: R & D Systems
MAB37701
102885-632EA 633.42 USD
102885-632
Anti-ANXA1 Mouse Monoclonal Antibody [clone: 686122]
Antibodies
Simple Western

Type: Primary
Antigen: ANXA1
Clonality: Monoclonal
Clone: 686122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR