Anti-ANXA1 Mouse Monoclonal Antibody [clone: 68616]

Supplier: R & D Systems
MAB3770
102885-630EA 655.11 USD
102885-630
Anti-ANXA1 Mouse Monoclonal Antibody [clone: 68616]
Antibodies
Type: Primary
Antigen: ANXA1
Clonality: Monoclonal
Clone: 68616
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR