Anti-CHUK Rat Monoclonal Antibody [clone: 829177]

Supplier: R & D Systems
MAB3768
102885-628EA 594.37 USD
102885-628
Anti-CHUK Rat Monoclonal Antibody [clone: 829177]
Antibodies
Type: Primary
Antigen: CHUK
Clonality: Monoclonal
Clone: 829177
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR