Anti-FRK Rat Monoclonal Antibody [clone: 393812]

Supplier: R & D Systems
MAB3766
220020-608EA 566.89 USD
220020-608
Anti-FRK Rat Monoclonal Antibody [clone: 393812]
Antibodies
Type: Primary
Antigen Symbol: FRK
Antigen Name: fyn-related Src family tyrosine kinase
Antigen Synonyms: GTK, PTK5, RAK
Clonality: Monoclonal
Clone: 393812
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR