Anti-RUNX3 Mouse Monoclonal Antibody [clone: 527327]

Supplier: R & D Systems
MAB3765
220020-607EA 633.42 USD
220020-607
Anti-RUNX3 Mouse Monoclonal Antibody [clone: 527327]
Antibodies
Simple Western

Type: Primary
Antigen: RUNX3
Clonality: Monoclonal
Clone: 527327
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR