Anti-CCL27 Mouse Monoclonal Antibody [clone: 124315]

Supplier: R & D Systems
MAB3762
220020-606EA 772.25 USD
220020-606
Anti-CCL27 Mouse Monoclonal Antibody [clone: 124315]
Antibodies
Type: Primary
Antigen: CCL27
Clonality: Monoclonal
Clone: 124315
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR