Anti-PLXNB1 Mouse Monoclonal Antibody [clone: 559819]

Supplier: R & D Systems
MAB37492
102885-626EA 577.02 USD
102885-626
Anti-PLXNB1 Mouse Monoclonal Antibody [clone: 559819]
Antibodies
Type: Primary
Antigen: PLXNB1
Clonality: Monoclonal
Clone: 559819
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR