Anti-PLXNB1 Mouse Monoclonal Antibody [clone: 439512]

Supplier: R & D Systems
MAB3749
220020-600EA 594.37 USD
220020-600
Anti-PLXNB1 Mouse Monoclonal Antibody [clone: 439512]
Antibodies
Type: Primary
Antigen: PLXNB1
Clonality: Monoclonal
Clone: 439512
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR