Anti-WNT11 Rat Monoclonal Antibody [clone: 310622]

Supplier: R & D Systems
MAB3746
102885-624EA 577.02 USD
102885-624
Anti-WNT11 Rat Monoclonal Antibody [clone: 310622]
Antibodies
Type: Primary
Antigen: WNT11
Clonality: Monoclonal
Clone: 310622
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR