Anti-COX1 Mouse Monoclonal Antibody [clone: 596002]

Supplier: R & D Systems
MAB37401
102885-622EA 633.42 USD
102885-622
Anti-COX1 Mouse Monoclonal Antibody [clone: 596002]
Antibodies
Type: Primary
Antigen: COX1
Clonality: Monoclonal
Clone: 596002
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR