Anti-COX1 Rat Monoclonal Antibody [clone: 345305]

Supplier: R & D Systems
MAB3740
220020-598EA 584.25 USD
220020-598
Anti-COX1 Rat Monoclonal Antibody [clone: 345305]
Antibodies
Type: Primary
Antigen: COX1
Clonality: Monoclonal
Clone: 345305
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR