Anti-NPPC Rat Monoclonal Antibody [clone: 347625]

Supplier: R & D Systems
MAB3739
220020-597EA 561.11 USD
220020-597
Anti-NPPC Rat Monoclonal Antibody [clone: 347625]
Antibodies
Type: Primary
Antigen: NPPC
Clonality: Monoclonal
Clone: 347625
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR