Anti-MC3R Rat Monoclonal Antibody [clone: 345404]

Supplier: R & D Systems
MAB3737
220020-596EA 577.02 USD
220020-596
Anti-MC3R Rat Monoclonal Antibody [clone: 345404]
Antibodies
Type: Primary
Antigen: MC3R
Clonality: Monoclonal
Clone: 345404
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR