Anti-TPSB2 Rat Monoclonal Antibody [clone: 286820]

Supplier: R & D Systems
MAB3736
220020-595EA 594.37 USD
220020-595
Anti-TPSB2 Rat Monoclonal Antibody [clone: 286820]
Antibodies
Type: Primary
Antigen: TPSB2
Clonality: Monoclonal
Clone: 286820
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR