Anti-NOTCH2 Mouse Monoclonal Antibody [clone: 602845]

Supplier: R & D Systems
MAB37351
102885-620EA 594.37 USD
102885-620
Anti-NOTCH2 Mouse Monoclonal Antibody [clone: 602845]
Antibodies
Type: Primary
Antigen: NOTCH2
Clonality: Monoclonal
Clone: 602845
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR