Anti-NOTCH2 Rat Monoclonal Antibody [clone: 411801]

Supplier: R & D Systems
MAB3735
220020-594EA 577.02 USD
220020-594
Anti-NOTCH2 Rat Monoclonal Antibody [clone: 411801]
Antibodies
Type: Primary
Antigen: NOTCH2
Clonality: Monoclonal
Clone: 411801
Conjugation: Unconjugated
Epitope: Intracellular Domain
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR