Anti-ADGRE5 Rat Monoclonal Antibody [clone: 58772]

Supplier: R & D Systems
MAB3734
102885-618EA 772.25 USD
102885-618
Anti-ADGRE5 Rat Monoclonal Antibody [clone: 58772]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: ADGRE5
Clonality: Monoclonal
Clone: 58772
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR