Anti-NTRK3 Mouse Monoclonal Antibody [clone: 75213]

Supplier: R & D Systems
MAB3731
220020-593EA 772.25 USD
220020-593
Anti-NTRK3 Mouse Monoclonal Antibody [clone: 75213]
Antibodies
Type: Primary
Antigen: NTRK3
Clonality: Monoclonal
Clone: 75213
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR