Anti-NR1I3 Rat Monoclonal Antibody [clone: 331930]

Supplier: R & D Systems
MAB3724
220020-591EA 577.02 USD
220020-591
Anti-NR1I3 Rat Monoclonal Antibody [clone: 331930]
Antibodies
Type: Primary
Antigen: NR1I3
Clonality: Monoclonal
Clone: 331930
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR