Anti-GDF3 Rat Monoclonal Antibody [clone: 356613]

Supplier: R & D Systems
MAB3719
220020-590EA 561.11 USD
220020-590
Anti-GDF3 Rat Monoclonal Antibody [clone: 356613]
Antibodies
Type: Primary
Antigen Symbol: GDF3 (Propeptide)
Antigen Name: growth differentiation factor 3
Antigen Synonyms: C78318, Gdf-3, Vgr-2, Vgr2, ecat9
Clonality: Monoclonal
Clone: 356613
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR