Anti-PLCG2 Mouse Monoclonal Antibody [clone: 346404]

Supplier: R & D Systems
MAB3716
220020-589EA 633.42 USD
220020-589
Anti-PLCG2 Mouse Monoclonal Antibody [clone: 346404]
Antibodies
Type: Primary
Antigen: PLCG2
Clonality: Monoclonal
Clone: 346404
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR