Anti-CCL23 Mouse Monoclonal Antibody [clone: 69523]

Supplier: R & D Systems
MAB3711
220020-587EA 763.57 USD
220020-587
Anti-CCL23 Mouse Monoclonal Antibody [clone: 69523]
Antibodies
Type: Primary
Antigen: CCL23
Clonality: Monoclonal
Clone: 69523
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR