Anti-PRSS3 Rat Monoclonal Antibody [clone: 420012]

Supplier: R & D Systems
MAB37101
220020-586EA 577.02 USD
220020-586
Anti-PRSS3 Rat Monoclonal Antibody [clone: 420012]
Antibodies
Type: Primary
Antigen: PRSS3
Clonality: Monoclonal
Clone: 420012
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR