Anti-ZAP70 Mouse Monoclonal Antibody [clone: 521626]

Supplier: R & D Systems
MAB3709
102885-318EA 577.02 USD
102885-318
Anti-ZAP70 Mouse Monoclonal Antibody [clone: 521626]
Antibodies
Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: 521626
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR