Anti-LCK Mouse Monoclonal Antibody [clone: 693010]

Supplier: R & D Systems
MAB37041
102885-316EA 577.02 USD
102885-316
Anti-LCK Mouse Monoclonal Antibody [clone: 693010]
Antibodies
Type: Primary
Antigen: LCK
Clonality: Monoclonal
Clone: 693010
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR