Anti-LCK Mouse Monoclonal Antibody [clone: 355713]

Supplier: R & D Systems
MAB3704
220020-583EA 594.37 USD
220020-583
Anti-LCK Mouse Monoclonal Antibody [clone: 355713]
Antibodies
Type: Primary
Antigen: LCK
Clonality: Monoclonal
Clone: 355713
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR