Anti-ORM1 Mouse Monoclonal Antibody [clone: 386131]

Supplier: R & D Systems
MAB3694
220020-581EA 594.37 USD
220020-581
Anti-ORM1 Mouse Monoclonal Antibody [clone: 386131]
Antibodies
Type: Primary
Antigen: ORM1
Clonality: Monoclonal
Clone: 386131
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR