Anti-EFNA4 Mouse Monoclonal Antibody [clone: 135305]

Supplier: R & D Systems
MAB369
220020-580EA 763.57 USD
220020-580
Anti-EFNA4 Mouse Monoclonal Antibody [clone: 135305]
Antibodies
Type: Primary
Antigen: EFNA4
Clonality: Monoclonal
Clone: 135305
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR