Anti-LRIG1 Rat Monoclonal Antibody [clone: 440710]

Supplier: R & D Systems
MAB3688
220020-579EA 561.11 USD
220020-579
Anti-LRIG1 Rat Monoclonal Antibody [clone: 440710]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: LRIG1
Clonality: Monoclonal
Clone: 440710
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR