Anti-LINGO2 Mouse Monoclonal Antibody [clone: 382007]

Supplier: R & D Systems
MAB36791
220020-578EA 594.37 USD
220020-578
Anti-LINGO2 Mouse Monoclonal Antibody [clone: 382007]
Antibodies
Type: Primary
Antigen: LINGO2
Clonality: Monoclonal
Clone: 382007
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR