Anti-C1RL Mouse Monoclonal Antibody [clone: 393608]

Supplier: R & D Systems
MAB3676
220020-575EA 561.11 USD
220020-575
Anti-C1RL Mouse Monoclonal Antibody [clone: 393608]
Antibodies
Type: Primary
Antigen: C1RL
Clonality: Monoclonal
Clone: 393608
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR