Anti-WNT9B Rat Monoclonal Antibody [clone: 406612]

Supplier: R & D Systems
MAB3669
220020-573EA 594.37 USD
220020-573
Anti-WNT9B Rat Monoclonal Antibody [clone: 406612]
Antibodies
Type: Primary
Antigen: WNT9B
Clonality: Monoclonal
Clone: 406612
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR