Anti-DSCAM Mouse Monoclonal Antibody [clone: 399212]

Supplier: R & D Systems
MAB36661
220020-572EA 577.02 USD
220020-572
Anti-DSCAM Mouse Monoclonal Antibody [clone: 399212]
Antibodies
Type: Primary
Antigen: DSCAM
Clonality: Monoclonal
Clone: 399212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR