Anti-CCL21 Mouse Monoclonal Antibody [clone: 54111]

Supplier: R & D Systems
MAB3661
220020-570EA 772.25 USD
220020-570
Anti-CCL21 Mouse Monoclonal Antibody [clone: 54111]
Antibodies
Type: Primary
Antigen: CCL21
Clonality: Monoclonal
Clone: 54111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR