Anti-TLR9 Mouse Monoclonal Antibody [clone: 229106]

Supplier: R & D Systems
MAB3658
220020-567EA 561.11 USD
220020-567
Anti-TLR9 Mouse Monoclonal Antibody [clone: 229106]
Antibodies
Type: Primary
Antigen: TLR9
Clonality: Monoclonal
Clone: 229106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR