Anti-MMP11 Mouse Monoclonal Antibody [clone: 135421]

Supplier: R & D Systems
MAB3657
220020-566EA 577.02 USD
220020-566
Anti-MMP11 Mouse Monoclonal Antibody [clone: 135421]
Antibodies
Type: Primary
Antigen: MMP11
Clonality: Monoclonal
Clone: 135421
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR