Anti-CLDN6 Mouse Monoclonal Antibody [clone: 342927]

Supplier: R & D Systems
MAB3656
220020-565EA 623.29 USD
220020-565
Anti-CLDN6 Mouse Monoclonal Antibody [clone: 342927]
Antibodies
Type: Primary
Antigen: CLDN6
Clonality: Monoclonal
Clone: 342927
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR