Anti-GPR15 Mouse Monoclonal Antibody [clone: 367902]

Supplier: R & D Systems
MAB3654
220020-563EA 647.88 USD
220020-563
Anti-GPR15 Mouse Monoclonal Antibody [clone: 367902]
Antibodies
Type: Primary
Antigen Symbol: GPR15
Antigen Name: G protein-coupled receptor 15
Antigen Synonyms: BOB
Clonality: Monoclonal
Clone: 367902
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR