Anti-SLC27A4 Mouse Monoclonal Antibody [clone: 342142]

Supplier: R & D Systems
MAB3650
220020-560EA 577.02 USD
220020-560
Anti-SLC27A4 Mouse Monoclonal Antibody [clone: 342142]
Antibodies
Type: Primary
Antigen: SLC27A4
Clonality: Monoclonal
Clone: 342142
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR