Anti-NOTCH1 Mouse Monoclonal Antibody [clone: 433802]

Supplier: R & D Systems
MAB3647
220020-556EA 584.25 USD
220020-556
Anti-NOTCH1 Mouse Monoclonal Antibody [clone: 433802]
Antibodies
Type: Primary
Antigen: NOTCH1
Clonality: Monoclonal
Clone: 433802
Conjugation: Unconjugated
Epitope: Intracellular Doman
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR