Anti-STAB2 Mouse Monoclonal Antibody [clone: 841101]

Supplier: R & D Systems
MAB3645
102885-610EA 594.37 USD
102885-610
Anti-STAB2 Mouse Monoclonal Antibody [clone: 841101]
Antibodies
Type: Primary
Antigen: STAB2
Clonality: Monoclonal
Clone: 841101
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR