Anti-CD48 Mouse Monoclonal Antibody [clone: 394620]

Supplier: R & D Systems
MAB36441
220020-553EA 561.11 USD
220020-553
Anti-CD48 Mouse Monoclonal Antibody [clone: 394620]
Antibodies
Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 394620
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR