Anti-CCL17 Mouse Monoclonal Antibody [clone: 54015]

Supplier: R & D Systems
MAB3641
220020-551EA 772.25 USD
220020-551
Anti-CCL17 Mouse Monoclonal Antibody [clone: 54015]
Antibodies
Type: Primary
Antigen: CCL17
Clonality: Monoclonal
Clone: 54015
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR