Anti-CCL15 Mouse Monoclonal Antibody [clone: 88119]

Supplier: R & D Systems
MAB3631
220020-549EA 772.25 USD
220020-549
Anti-CCL15 Mouse Monoclonal Antibody [clone: 88119]
Antibodies
Type: Primary
Antigen: CCL15
Clonality: Monoclonal
Clone: 88119
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR