Anti-CCL15 Mouse Monoclonal Antibody [clone: 52130]

Supplier: R & D Systems
MAB363
220020-547EA 772.25 USD
220020-547
Anti-CCL15 Mouse Monoclonal Antibody [clone: 52130]
Antibodies
Type: Primary
Antigen: CCL15
Clonality: Monoclonal
Clone: 52130
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR