Anti-PECAM1 Rat Monoclonal Antibody [clone: 693102]

Supplier: R & D Systems
MAB3628
102885-608EA 594.37 USD
102885-608
Anti-PECAM1 Rat Monoclonal Antibody [clone: 693102]
Antibodies
Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 693102
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR