Anti-IL33 Rat Monoclonal Antibody [clone: 390412]

Supplier: R & D Systems
MAB3625
220020-545EA 642.09 USD
220020-545
Anti-IL33 Rat Monoclonal Antibody [clone: 390412]
Antibodies
Type: Primary
Antigen: IL33
Clonality: Monoclonal
Clone: 390412
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR