Anti-CMKLR1 Mouse Monoclonal Antibody [clone: 84939]

Supplier: R & D Systems
MAB362
220020-544EA 763.57 USD
220020-544
Anti-CMKLR1 Mouse Monoclonal Antibody [clone: 84939]
Antibodies
Type: Primary
Antigen: CMKLR1
Clonality: Monoclonal
Clone: 84939
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR