Anti-FOXJ1 Rat Monoclonal Antibody [clone: 407003]

Supplier: R & D Systems
MAB3619
220020-543EA 594.37 USD
220020-543
Anti-FOXJ1 Rat Monoclonal Antibody [clone: 407003]
Antibodies
Type: Primary
Antigen Symbol: FOXJ1
Antigen Name: forkhead box J1
Antigen Synonyms: FKHL13, HFH-4, HFH4
Clonality: Monoclonal
Clone: 407003
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR