Anti-PYGO1 Mouse Monoclonal Antibody [clone: 466023]

Supplier: R & D Systems
MAB3617
220020-542EA 577.02 USD
220020-542
Anti-PYGO1 Mouse Monoclonal Antibody [clone: 466023]
Antibodies
Type: Primary
Antigen: PYGO1
Clonality: Monoclonal
Clone: 466023
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR